missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 6-nitroindole-2-carboxylate, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
Description
Ethyl 6-nitroindole-2-carboxylate used as a intermediate in pharmaceutical and chemical synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethyl 6-nitroindole-2-carboxylate used as a intermediate in pharmaceutical and chemical synthesis.
Solubility
Slightly soluble in water.
Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Ethyl 6-nitroindole-2-carboxylate used as a intermediate in pharmaceutical and chemical synthesis.
Solubility
Slightly soluble in water.
Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.

Specifications
Specifications
Color | Yellow |
Quantity | 250 mg |
Solubility Information | Slightly soluble in water. |
Formula Weight | 234.21 |
Percent Purity | 97% |
Physical Form | Solid |
Chemical Name or Material | Ethyl 6-nitroindole-2-carboxylate |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ethyl 6-nitroindole-2-carboxylate, 97%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction