missing translation for 'onlineSavingsMsg'
Learn More

Ethyl cis-2-amino-1-cyclohexanecarboxylate hydrochloride, 95%

Product Code. p-127485 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
2.5 g
Packaging:
Glass Bottle
Unit Size:
1g
2.5g
This item is not returnable. View return policy

Product Code. 10781711

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1127-99-7 | C9H17NO2·HCl | 207.7 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1127-99-7
Molecular Formula C9H17NO2·HCl
Molecular Weight (g/mol) 207.7
MDL Number MFCD00144280
InChI Key XMQSOBPCWYVZSW-WLYNEOFISA-N
Synonym ethyl cis-2-amino-1-cyclohexanecarboxylate hydrochloride, cis-2-amino-cyclohexanecarboxylic acid ethyl ester hydrochloride, ethyl 1r,2s-2-aminocyclohexane-1-carboxylate hydrochloride, cis-ethyl 2-aminocyclohexanecarboxylate hydrochloride, 1r,2s-ethyl 2-aminocyclohexanecarboxylate hydrochloride, cyclohexanecarboxylic acid, 2-amino-, ethyl ester, hydrochloride, cis, ethyl cis-2-amino-1-cyclohexanecarboxylate hcl salt, ethyl cis-2-aminocyclohexanecarboxylate hydrochloride, ethyl 2s,1r-2-aminocyclohexanecarboxylate, chloride
PubChem CID 2724641
IUPAC Name ethyl (1R,2S)-2-aminocyclohexane-1-carboxylate;hydrochloride
SMILES CCOC(=O)C1CCCCC1N.Cl

Specifications

Melting Point 128°C to 135°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass Bottle
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in methanol
Quantity 1 g
Formula Weight 207.7
Percent Purity 95%
Physical Form Crystalline Powder or Chunks
Chemical Name or Material Ethyl cis-2-amino-1-cyclohexanecarboxylate hydrochloride, 95%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.