Learn More
Etoricoxib-d4, MedChemExpress
MedChemExpress Etoricoxib D4 (MK-0663 D4) is a deuterium labeled Etoricoxib. Etoricoxib is a non steroidal anti-inflammatory agent, acting as a selective and orally active COX-2 inhibitor, with IC50s of 1.1 μM and 116 μM for COX-2 and COX-1 in human whole blood.
Brand:  Medchem Express HY-15321S
Encompass_Preferred
 View more versions of this product
Product Code. 16481499
  • 251.10 € / 1mg
Please to purchase this item. Need a web account? Register with us today!
Explore more special offers
Description
Description
Etoricoxib D4 (MK-0663 D4) is a deuterium labeled Etoricoxib. Etoricoxib is a non steroidal anti-inflammatory agent, acting as a selective and orally active COX-2 inhibitor, with IC50s of 1.1 μM and 116 μM for COX-2 and COX-1 in human whole blood.
Specifications
Specifications
Etoricoxib-d4
1 mg
Research
MK-0663-d4 L-791456-d4
ClC(C=N1)=CC(C2=C([2H])C([2H])=C(S(C)(=O)=O)C([2H])=C2[2H])=C1C3=CN=C(C)C=C3
362.87
Research
1131345-14-6
Light Nude
C18H11D4ClN2O2S
Powder
362.87
99.35%