missing translation for 'onlineSavingsMsg'
Learn More

IKA™ EUROSTAR 400 Control, Overhead Stirrer
GREENER_CHOICE

Powerful overhead stirrer for highly viscous fluids and intensive mixing for large volumes.

Brand:  IKA™ 0004214100

Product Code. 15710728

  • 5295.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Multilingual TFT display
  • Programmable functions
  • Integrated temperature measurement
  • Interval operation
  • Timer function
  • Adjustable safety circuit
  • Locked function
  • Infinitely adjustable speed
  • Push-through agitator shafts
  • Overload protection
  • Short-term overload operation
  • Slim casing
  • Quiet operation
  • Error code display
TRUSTED_SUSTAINABILITY
Specifications

Specifications

0/6 to 2000 rpm
TFT
226 w
230/115/100 V, 50/60 HZ, 226 W
3 to 16 mm
EUROSTAR 400 Control
400 Ncm
8.2 kg
150 L
100/115/230 V
114 x 320 x 268 mm
50/60 Hz
Alu-Cast Coating/Thermoplastic Polymer
Brushless DC
100,000 mPas
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
IKA™ EUROSTAR 400 Control, Overhead Stirrer > 150 L Capacity

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.