missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Eutech PH 1710 pH Bench Meter
Eutech PH 1710 pH bench meters offer a basic, cost-effective testing option for measuring pH or relative mV (RmV) with temperature. Each Eutech 1710 pH meter features a large display with backlight, easy operation and meter-integrated electrode holder.
Brand: Thermo Scientific™ EBPH171003
This item is not returnable.
View return policy
Description
Eutech PH 1710 pH bench meters offer a basic, cost-effective testing option for measuring pH or relative mV (RmV) with temperature. Each Eutech 1710 pH meter features a large display with backlight, easy operation and meter-integrated electrode holder.
Select Eutech PH 1710 pH bench meters for a basic, cost-effective option for your pH and relative mV (RmV) testing needs. Eutech 1710 pH meters measure pH or RmV with temperature. Each Eutech 1710 pH meter features a large display with backlight, easy operation and meter-integrated electrode holder.Specifications
Eutech PH 1710 pH/mV bench meter low-maintenance kit | |
±0.3°C | |
±0.01 pH ± 1 LSD | |
Computer or printer (STARA-106) | |
pH with mV and Temperature, Relative mV with mV and Temperature | |
-2 to 18 | |
1 to 5 points | |
±2000.0 mV with ±250 mV offset | |
1 point RmV | |
0.1°C | |
1 point temperature offset calibration | |
Yes | |
500 data sets with time and date | |
Calibration reminder alarm | |
CE, TUV 3-1, FCC Class A | |
Universal 100-240 V 50/60 Hz power supply with twist-and-lock meter connection and interchangeable wall plugs for US/Japan, Euro, UK/Singapore, Australia/New Zealand and China outlets | |
700 g | |
3 year meter warranty |
Meter with electrode stand, PC cable, universal power adapter | |
±0.2 or ±0.05%, whichever is greater | |
Transfer single data point or all data points to computer or printer | |
For Samples Containing TRIS Buffers, Proteins, Sulfides or Halides; Where a Gel-Filled pH Electrode That is Low-Maintenance and Easy to Operate is Desirable | |
pH, RmV and temperature | |
0.1, 0.01 pH | |
±2000.0 mV | |
0.1 mV | |
-5.0°C to +105.0°C | |
Manual or automatic with ATC probe | |
5 in. backlit LCD | |
Non-volatile | |
Automatic with Auto-Read and Timed read modes, manual with Continuous read mode | |
BNC for pH or RmV and phono plug for ATC | |
IP54 | |
198.3 x 155.2 x 61.7 mm (7.81 x 6.11 x 2.43 in.) | |
1.54 lb. |