Learn More
EVOS™ EVOS™ Vessel Holder, ibidi 50 mm Petri dishes
For flasks, trays and petri dishes
Brand: EVOS™ AMEPVH014
148.20 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.
This item is not returnable.
View return policy
Description
Broad range of holders helps maximize the flexibility of the EVOS and Micromaster™ inverted microscopes.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Vessel Holder | |
EVOS™ XL Core Imaging System, EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System | |
2 50 mm ibidi Petri Dishes |
EVOS | |
Ibidi 50 mm Petri dishes | |
1 each |