Learn More
Memmert™ Extended Overtemperature Protection for UF55
Product Code. 17539425 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
UF55 Oven
Unit Size:
Each
Product Code. 17539425
Brand: Memmert™ UF55A6
Only null left
Add to basket
Add to basket
Extended overtemperature protection with integrated Pt100 sensor for independent temperature monitoring.
  • Accessory
  • With integrated Pt100 sensor
  • For models with SingleDISPLAY
Specifications
For Use With (Equipment) UF55 Oven
Includes Integrated Pt100 Sensor