Learn More
Memmert™ Extension of DAkkS Factory Calibration for SN160
Extension of DAkkS factory calibration by an additional freely selectable temperature for Memmert Sterilizers.
Brand: Memmert™ SN160E39697
This item is not returnable.
View return policy
Description
- Extension of DAkkS calibration at Memmert factory by an additional freely selectable temperature value according to method C (DKD-R 5-7)
Specifications
SN160 Sterilizer | |
Factory Calibration Extension |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ Extension of DAkkS Factory Calibration for SN160 > For Use With: SN160
Spot an opportunity for improvement?Share a Content Correction