Learn More
Memmert™ Extension of DAkkS Factory Calibration for SN260
Extension of DAkkS factory calibration by an additional freely selectable temperature for Memmert Sterilizers.
Brand:  Memmert™ SN260E39697
Product Code. 17598545
  • 979.00 € / Each
Please to purchase this item. Need a web account? Register with us today!
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • Extension of DAkkS calibration at Memmert factory by an additional freely selectable temperature value according to method C (DKD-R 5-7)
Specifications
Specifications
SN260 Sterilizer
Factory Calibration Extension
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ Extension of DAkkS Factory Calibration for SN260 > For Use With: SN260