Learn More
Memmert™ Extension of DAkkS Factory Calibration for UF55
Extension of DAkkS factory calibration by an additional freely selectable temperature for Memmert Ovens.
Brand: Memmert™ UF55E39697
This item is not returnable.
View return policy
Description
- Extension of DAkkS calibration at Memmert factory by an additional freely selectable temperature value according to method C (DKD-R 5-7)
Specifications
UF55 Oven | |
Factory Calibration Extension |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ Extension of DAkkS Factory Calibration for UF55 > For Use With: UF55
Spot an opportunity for improvement?Share a Content Correction