Learn More
Memmert™ Extension of DAkkS Factory Calibration for UF75
Product Code. 17519665 Shop All Memmert Products
Click to view available options
Unit Size:
Each
Product Code. 17519665
Brand: Memmert™ UF75E39697
Please to purchase this item. Need a web account? Register with us today!
Extension of DAkkS factory calibration by an additional freely selectable temperature for Memmert Ovens.
  • Extension of DAkkS calibration at Memmert factory by an additional freely selectable temperature value according to method C (DKD-R 5-7)
Specifications
Product Type Factory Calibration Extension