Learn More
Lenz Laborglasintrumente Extractors for Specific Heavy Solvents
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 05433058
 View more versions of this product
Product Code. 10191421
  • 637.00 € / Each
Estimated Shipment: 04-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Extractor head with insert filter (frit, porosity: 0), with PTFE valve stopcock
  • Dimroth condenser with thread GL 14
  • Round bottom flask
  • DURAN™
Specifications
Specifications
500 mL
0
NS 60/46