Learn More
Plum™ Eyewash Box
Wall-mounted eyewash boxes made in maintenance-free material for light and quick eye injury first aid
Brand:  Plum™ 4650
Product Code. 10323513
  • 146.00 € / Each
Estimated Shipment: 13-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Dust-tight wall stations in high-impact polystyrene
  • Visible and readily available first aid
  • Quick and efficient eyewash
  • Simple and quickly mounted on the wall close to the workplace
  • May be mounted right where the need for immediate rinsing will be
  • Supplied with clear pictograms and mirror
  • Particularly suited for dusty and dirty production areas
  • Suited for mobile work places
  • Specifications
    Specifications
    110 mm
    280 mm
    2 x 500 mL Plum Eyewash
    230 mm