Learn More
F4/80 Antigen Rat, FITC, Clone: BM8.1, enQuireBio™
Rat Monoclonal Antibody
Brand:  EnquireBio QAB68-F-500ug
 View more versions of this product
Product Code. 15952358
  • 377.00 € / 500µg
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
Specifications
Specifications
F4/80 Antigen
Monoclonal
0.5mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2
Adgre1
Protein A/G Purified
RUO
13733
IgG2b κ
Flow Cytometry
BM8.1
FITC
XP_006523664
Rat
500 μg
Primary
Store at 2-4°C.