Learn More
Memmert™ Factory Calibration (3 Temperatures) for UF750
Calibration at Memmert factory for three temperature value.
Brand: Memmert™ UF750D00128
This item is not returnable.
View return policy
Description
- For three temperature values: 100°C, 160°C, and 220°C
Specifications
UF750 Oven | |
Measuring point chamber center | |
Certificate for 3 temperatures | |
Standard Works Calibration Certificate |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ Factory Calibration (3 Temperatures) for UF750 > For Use With: UF750
Spot an opportunity for improvement?Share a Content Correction