Learn More
Memmert™ Factory Calibration (3 Temperatures) for UN160plus
Calibration at Memmert factory for three temperature value.
Brand: Memmert™ UN160plusD00128
This item is not returnable.
View return policy
Description
- For three temperature values: 100°C, 160°C, and 220°C
Specifications
UN160plus Oven | |
Measuring point chamber center | |
Certificate for 3 temperatures | |
Standard Works Calibration Certificate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ Factory Calibration (3 Temperatures) for UN160plus > For Use With: UN160plus
Spot an opportunity for improvement?Share a Content Correction