Learn More
Falcon™ HTS 24-Well Multiwell Permeable Support System
Save time and labor in HTS protocols—grow, handle, and analyze 24 samples simultaneously. Falcon™ HTS Multiwell Insert System features a generous, automation-compatible sampling port which allows sampling above and below membranes.
Brand:  Falcon™ 351180
 View more versions of this product
Product Code. 10050861
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Includes: 24-well insert plate, 24-well plate or single-well feeder tray, and lid
Description
Description
Falcon 24 Multiwell Insert Systems are designed to automate many commonly used cell-based assays for drug discovery. Available in a choice of membrane pore sizes, 24 Multiwell inserts have been successfully used for a variety of applications including permeability studies for oral bioavailability (e.g., Caco-2 cells), chemotaxis, cell migration and invasion assays. These insert systems offer all the benefits of Falcon Individual Cell Culture Inserts in an automation-friendly format that is compatible with most robots and fluid handlers. The Falcon 24 Multiwell Insert Plate is a single unit that is compatible with all Falcon 24 well plates and Feeder Tray.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Yes
0.3 cm2
Sterile
24
24-well insert plate, single well feeder tray and lid
Each
Clear
PETE
Multi-well Insert System
Excellent for differentiation, toxicology, migration and invasion studies
1 μm
Polyethylene Terephthalate