Learn More
Falcon™ HTS 24-Well Multiwell Permeable Support System
Save time and labor in HTS protocols—grow, handle, and analyze 24 samples simultaneously. Falcon™ HTS Multiwell Insert System features a generous, automation-compatible sampling port which allows sampling above and below membranes.
Brand: Falcon™ 351182
Includes: 24-well insert plate, 24-well plate or single-well feeder tray, and lid
Description
Falcon 24 Multiwell Insert Systems are designed to automate many commonly used cell-based assays for drug discovery. Available in a choice of membrane pore sizes, 24 Multiwell inserts have been successfully used for a variety of applications including permeability studies for oral bioavailability (e.g., Caco-2 cells), chemotaxis, cell migration and invasion assays. These insert systems offer all the benefits of Falcon Individual Cell Culture Inserts in an automation-friendly format that is compatible with most robots and fluid handlers. The Falcon 24 Multiwell Insert Plate is a single unit that is compatible with all Falcon 24 well plates and Feeder Tray.
Specifications
Yes | |
0.3 cm2 | |
Sterile | |
24 | |
24-well insert plate, 24-well plate and lid | |
Each |
Clear | |
Polystyrene | |
Multi-well Insert System | |
Excellent for differentiation, toxicology, migration and invasion studies | |
3 μm | |
Polyethylene Terephthalate |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.