Learn More
Falcon™ HTS 24-Well Multiwell Permeable Support System
Save time and labor in HTS protocols—grow, handle, and analyze 24 samples simultaneously. Falcon™ HTS Multiwell Insert System features a generous, automation-compatible sampling port which allows sampling above and below membranes.
Brand:  Falcon™ 351182
 View more versions of this product
Product Code. 10438642
Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen
View alternative products »
Explore more special offers
Includes: 24-well insert plate, 24-well plate or single-well feeder tray, and lid
Description
Description
Falcon 24 Multiwell Insert Systems are designed to automate many commonly used cell-based assays for drug discovery. Available in a choice of membrane pore sizes, 24 Multiwell inserts have been successfully used for a variety of applications including permeability studies for oral bioavailability (e.g., Caco-2 cells), chemotaxis, cell migration and invasion assays. These insert systems offer all the benefits of Falcon Individual Cell Culture Inserts in an automation-friendly format that is compatible with most robots and fluid handlers. The Falcon 24 Multiwell Insert Plate is a single unit that is compatible with all Falcon 24 well plates and Feeder Tray.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Yes
0.3 cm2
Sterile
24
24-well insert plate, 24-well plate and lid
Each
Clear
Polystyrene
Multi-well Insert System
Excellent for differentiation, toxicology, migration and invasion studies
3 μm
Polyethylene Terephthalate