Learn More
Instech Laboratories™ Feeding tube, mouse oral gavage, polyurethane, 16ga x 38mm, sterile
Brand: Instech Laboratories™ FTPU-16-38-50
Description
These specialty tubes feature a larger inner diameter for easier delivery of viscous compounds and a straight tip that wipes clean to help in cases where aspiration of compounds would be hazardous to the animal. Availablein two sizes: 13ga for rats and 16ga for mice.
- Plastic feeding tubes, 16ga x 38mm
- For mouse oral gavage
- Designed to minimise trauma to oesophagus
- Polyurethane tube with rounded tip
- Large internal diameter for viscous compounds
- Straight tube wipes clean to prevent aspiration of compounds
- Disposable
- Supplied as 10 sterile pouches of 5 tubes
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.