Learn More
Instech Laboratories™ Feeding tube, mouse oral gavage, polyurethane, 16ga x 38mm, sterile
Brand: Instech Laboratories™ FTPU-16-38
This item is not returnable.
View return policy
Description
These specialty tubes feature a larger inner diameter for easier delivery of viscous compounds and a straight tip that wipes clean to help in cases where aspiration of compounds would be hazardous to the animal. Availablein two sizes: 13ga for rats and 16ga for mice.- Plastic feeding tubes, 16ga x 38mm
- For mouse oral gavage
- Designed to minimise trauma to oesophagus
- Polyurethane tube with rounded tip
- Large internal diameter for viscous compounds
- Straight tube wipes clean to prevent aspiration of compounds
- Disposable
- Supplied as 50 sterile pouches of 5 tubes