Learn More
Thermo Scientific™ Fiberboard Box Dividers for Ultra-Low Temperature and Cryogenic Freezers
Product Code. 10059500 Shop All Thermo Scientific Products
Encompass
Click to view available options
Holds:
25 (5 x 5 Grid)
49 (7 x 7 Grid)
64 (8 x 8 Grid)
81 (9 x 9 Grid)
Quantity:
12 Pack
Each
Unit Size:
Each
Pack of 12
This item is not returnable. View return policy
Product Code. 10059500
Brand: Thermo Scientific™ 6212
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
Organize vial storage within cryo boxes with these fiberboard box dividers.
Organize vial storage with Thermo Scientific™ Fiberboard Box Dividers for Ultra-Low Temperature and Cryogenic Freezers.
  • Use with fiberboard storage boxes to divide box into grids for vial storage
  • Box dividers for cold storage application available in 7 x 7 , 8 x 8, 9 x 9 or 10 x 10 grids
Specifications
Quantity 12 Pack
Holds 81 (9 x 9 Grid)