Learn More
Finely Ground Edge Glass Microscopic Slides
Packed with tissue to protect glass surfaces. Scientific Glass Labs™ Finely Ground Edge Glass Microscope Slides are best used with large sections.
Brand:  Fisherbrand MNK-130-060P
Encompass
 View more versions of this product
Product Code. 10221722
  • 62.85 € / Pack of 50
Estimated Shipment: 28-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
With finely ground edges. Packed with tissue to protect glass surfaces. For use with large sections.
Specifications
Specifications
76 mm
Finely Ground
50 Pack
52 mm
1.2 to 1.5 mm
76 x 52 mm