Learn More
Thermo Scientific™ Finger Tite Connectors for Agilent Instruments
Thermo Scientific™ Finger Tite Connectors for Agilent Instruments offer easier fit, reliabililty and leak-free connections.
Brand: Thermo Scientific™ 290SA132
361.19 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Thermo Scientific™ Finger Tite Connectors for Agilent Instruments offer easier fit, reliabililty and leak-free connections.
Advantages over conventional graphite/polymer ferrules:
- Perfect metal-to-metal seal eliminates contamination and gives lower air/water background
- No need to retighten ferrules, as they expand and contract with the fitting over GC operating temperatures
- Easy to handle with the nut touchable even with a hot injector/detector
- No tools required
Specifications
Split/Splitless and FID Starter Kit | |
Injector/Detector Starter Kit; 1/Pk. | |
TRACE 1300 Series GC and Agilent GC Instruments | |
1/Pk. |