Learn More
DWK Life Sciences Fiolax™ Borosilicate Test Tube, with beaded rim
Product Code. 10078190 Shop All DWK Life Sciences Products
Click to view available options
Capacity (Metric):
2 mL
4 mL
6 mL
6.5 mL
9 mL
16 mL
20 mL
25 mL
35 mL
39 mL
45 mL
60 mL
80 mL
110 mL
Unit Size:
Pack of 100
Pack of 50
Product Code. 10078190
Brand: DWK Life Sciences 261101302
Only null left
Add to basket
Add to basket
Test tubes are universal cylindrical laboratory glassware with a round bottom that are used e.g. for chemical reactions, pretreatments or for storing small amounts of liquid.
Transferring substances and liquids into other states of aggregation and thus initiating further reactions is very important for the daily work in the laboratory. A wide variety of vessels, apparatus and equipment are used for these work. Reaction controls, test series and preliminary samples are usually carried out in test tubes due to the small amounts of substance removed.
  • Thin-walled and therefore resistant to rapid temperature changes or localized heating
  • Easier pouring due to beaded rim
  • Fired-on, highly durable white ceramic
TRUSTED_SUSTAINABILITY
Specifications
Description 0.4 to 0.5 mm thickness
Capacity (Metric) 16 mL
Material Fiolax
For Use With Heating of small amounts; Pretreatments; Storage of small amounts of liquid
Quantity 100/Pk.
Autoclavable Autoclavable
Diameter (Metric) 14 mm