Learn More
Fisherbrand™ Air-Powered Analog Overhead Stirrers
Variable speed and explosion-proof stirrers ideal for safely mixing all types of solvents, lacquers and volatile chemicals without the possiblity of spark ignition.
Brand:  Fisherbrand™ 37020-9015
 View more versions of this product
Product Code. 15122997
  • 1175.00 € / Each
Estimated Shipment: 13-06-2025
to see stock.
Explore more special offers
Add to basket
Includes:
  • A 9/16 in. dia. x 10 in. L support rod
  • Four ft. air hose with double fittings for connection to air supply
Description
Description
  • Needle valve allows variation in mixing speed
  • Special muffler ensures quieter operation
  • Reversible models have motors which can be set to rotate in either direction by connecting the air supply line to the right port for counter-clockwise rotation or the left port for clockwise rotation
  • Can be used with flammable chemicals for explosion proof mixing
Order Info
Shaft, propeller, stand and clamp available separately
Specifications
Specifications
1000 rpm
Analog
0.37 in.
10.5 in.
12 in.
0.5
10 L x 0.56 in. dia. support rod, 4 ft. air hose with double fittings for connection to air supply
30 to 80 psi
Air Powered Overhead Stirrer
6 lb.
20 L
0/1 V
0.95 cm
27 cm
30 cm
Safely mixing all types of solvents, lacquers and volatile chemicals
Steel Aluminum
294
10,000 mPas
2.7 kg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Air-Powered Analog Overhead Stirrers > Max. volume: 20L; Speed: 1000rpm; Max. torque: 294 Ncm;