accessibility menu, dialog, popup
Test
chevron-down
Select Language
English
German
Search
Order Status
Quick Order
Support
Sign In
chevron-down
chevron-down
Learn More
Home
Products
pH and Electrochemistry
pH and Electrochemistry Meter Accessories
Multiparameter Meter Accessories
12890653
Print
Fisherbrand™ RS232 Thermal Printer for accumet™ AB and XL Series Benchtop Meters: Paper Rolls
Printer Paper Rolls, Pack of 2
For use with RS-232 thermal printer
Brand:
Fisherbrand™ 13-637-691
Product Code.
12890653
47.00 €
/ Each
Quantity
Estimated Shipment: 29-01-2025
Log in
to see stock.
Explore more special offers
Add to basket
Description
Specifications
Product Suggestions
Videos
SDS
Documents
Certificates
Promotions
Description
Description
For use with RS-232 thermal printer
Paper Width: 58.4mm (2.3 in.)
Specifications
Specifications
Type
Thermal Printer
Width (English)
2.3 in.
Width (Metric)
0.06 m