Learn More
Fisherbrand™ Traceable™ Logger-Trac™ Temp. RH and LN2 Datalogger Cable
Shop All Fisherbrand Products
Click to view available options
Length (Metric):
1.5 m
Unit Size:
Each
Spezifikation
Spezifikation
| Type | Cable |
| Certifications/Compliance | RoHS compliant |
| For Use With (Application) | Transfer recorded information in dataloggers to computer |
| For Use With (Equipment) | Fisherbrand Traceable Logger-Trac Temperature/RH and LN2 Datalogger |
| Length (English) | 5 ft. |
| Length (Metric) | 1.5 m |