Learn More
Fisherbrand™ Disposable Borosilicate Glass Tubes with Threaded End
Product Code. 11557423 Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
8 mL
12 mL
16 mL
20 mL
25 mL
30 mL
36 mL
Length (Metric):
100 mm
125 mm
150 mm
Label Type:
Marking Spot
None
Thread Size:
13-415
15-415
18-415
Unit Size:
Pack of 1000
Pack of 500
Product Code. 11557423
Please to purchase this item. Need a web account? Register with us today!
Available with or without marking spot
  • Round bottom
  • Without caps
Order Info
Supplied in shrink-wrapped shelf trays of 500
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ASTM E438, Type I, Class B
Capacity (Metric) 36 mL
Length (Metric) 150 mm
Material Borosilicate Glass
Disposable Single-use
For Use With For tissue culture work and general bacteriological use
Label Type Marking Spot
Packaging Tray
Thread Size 18-415
Sterility Non-sterile
Diameter (Metric) 20 mm
Closure Type Not Included
Product Type Disposable Culture Tube