Learn More
Fisherbrand™ Electroporation Cuvettes Plus™
Disposable, color-coded cuvets fit most electroporators. Sterilized by gamma irradiation and supplied individually wrapped.
Brand:  Fisherbrand™ 450150
212.69 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.
 View more versions of this product
Product Code. 15532423
  • 305.00 € / Pack of 50
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
Disposable, color-coded cuvets are compatible with most electroporators.
  • Clear, medical grade polycarbonate for high visibility
  • Polished aluminum lowers arcing frequency
  • Round cap allows single-handed removal
  • FB102 has imbedded aluminum electrodes
Specifications
Specifications
Sterile
90 μL
Imbedded aluminum electrodes
Electroporator
Electroporation Cuvet
Polycarbonate
1 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24155
    Expires2025-04-30
    Days
    25
    Hours
    6
    Minutes
    10
    Seconds
    42
    Hot Deal
    *Discount will apply to qualifying orders received by Fisher Scientific between 01/03/2025 and 31/03/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions See Full Promo Details
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Electroporation Cuvettes Plus™ > 1mm gap; 90μL