Learn More
IKA™ Fixing Clips for Shakers
Brand:  IKA™ 0001234300
 View more versions of this product
Product Code. 10079400
  • 32.50 € / Each
Estimated Shipment: 27-03-2025
to see stock.
Explore more special offers
Add to basket
Specifications
25 mL Flask
Fixing Clip
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
IKA™ Fixing Clips for Shakers > For Use With: 25mL Flask