Learn More
Invitrogen™ FL - SRC 3-1 Peptide, 100μM
Contains known interaction motifs and labeled with fluorescein. Invitrogen™ Fluorescein-SRC 3-1 Peptide, 100μM is matched to complement the LanthaScreen™ TR-FRET Nuclear Receptor Coregulator Assays to enable primary or secondary screening of nuclear receptor agonists and antagonists.
Brand: Invitrogen™ PV4590
Description
Ligand binding to nuclear receptors causes conformational changes in the receptor, resulting in a cascade of events, including dissociation of repressor proteins, association of coactivator proteins, and assembly of pol II and other transcriptional factors for activation of target genes. TR-FRET based assays can be developed using the LanthaScreen™ panel of fluorescein-labeled coregulator peptides to investigate conformational changes of nuclear receptors upon ligand binding, either by determining the affinity of ligand-bound receptor for different coregulator peptides, or by identifying additional agonists or antagonists via displacement or recruitment of a specific coregulator peptide.
In the LanthaScreen™ TR-FRET RXR beta Coactivator Assay, a terbium (Tb)-labeled anti-GST antibody is used to indirectly label GSTtagged RXR beta protein. An agonist (9-cis Retinoic Acid) added to the receptor which, upon ligand binding, causes a conformational change resulting in recruitment of the Fl-D22 coactivator peptide. The binding of the fluorescent coregulator peptide to RXR beta causes an increase in the TR-FRET emission ratio.
Specifications
Store in freezer (-5 to -30°C). | |
Classic Dyes | |
100 μL | |
Dry Ice |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1899812J | Certificate of Analysis | 21/02/2025 | PV4590 |
1899812H | Certificate of Analysis | 19/05/2023 | PV4590 |
1899812G | Certificate of Analysis | 09/10/2021 | PV4590 |
1899812F | Certificate of Analysis | 23/02/2020 | PV4590 |
1899812E | Certificate of Analysis | 07/08/2019 | PV4590 |