Learn More
Grant Instruments™ Flask Tray for Grant Shaking Water Baths
Include threaded holes to accept flask clamps or holder for deep well plates (=2ml). Grant Instruments™ Flask Tray for Grant Shaking Water Baths are compatible with SR plate holders and racks or can be used alone to accommodate bags and miscellaneous vessels.
321.00 € - 349.00 €
Specifications
Capacity (Metric) 25 to 1000 mL
Type Flask Tray
View More Specs
Products 2
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
15345822
View Documents
Grant Instruments™
TF12
LSB12
321.00 €
Each
In Stock
to see stock.
Add to basket
15375822
View Documents
Grant Instruments™
TF18
LSB18
349.00 €
Each
In Stock
to see stock.
Add to basket
Description
Description
Include threaded holes to accept flask clamps or holder for deep well plates (=2ml). Grant Instruments™ Flask Tray for Grant Shaking Water Baths are compatible with SR plate holders and racks or can be used alone to accommodate bags and miscellaneous vessels.
  • Accommodates flasks from 25 to 1000mL
  • For use with the followng Shaking Water Bath models:
    • LSB12
    • LSB18
    • OLS26
  • Compatible with SH plate holders or SR racks
  • TF26 Test tube tray also compatible with SR racks
  • Can be used alone to accommodate bags and miscellaneous vessels.
Specifications
Specifications
25 to 1000 mL
Flask Tray
Videos
SDS
Documents
Documents
Product Certifications