Learn More
Memmert™ Flexible Pt100 Temperature Sensor for UF1060
Flexible temperature sensor for positioning in chamber or in load.
Brand: Memmert™ UF1060H8
Description
- Positioned flexibly in chamber or load
- For local temperature measurement (up to 1 additional sensor is possible)
- The measured temperature will be indicated on the display, recorded in the integral data store, and can be documented via the AtmoCONTROL software
- For Memmert Ovens
Specifications
UF1060 Oven | |
Temperature Sensor |