Learn More
Memmert™ Flexible Pt100 Temperature Sensor for UN260plus
Flexible temperature sensor for positioning in chamber or in load.
Brand: Memmert™ UN260plusH8
This item is not returnable.
View return policy
Description
- Positioned flexibly in chamber or load
- For local temperature measurement (up to 1 additional sensor is possible)
- The measured temperature will be indicated on the display, recorded in the integral data store, and can be documented via the AtmoCONTROL software
- For Memmert Ovens
Specifications
UN260plus Oven | |
Temperature Sensor |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ Flexible Pt100 Temperature Sensor for UN260plus > For Use With: UN260plus
Spot an opportunity for improvement?Share a Content Correction