missing translation for 'onlineSavingsMsg'
Learn More

FLI1 Antibody (FLI1/4371R), DyLight 350, Novus Biologicals™

Rabbit Monoclonal Antibody

Brand:  Novus Biologicals NBP3-21020UV

Product Code. 18517849

  • 729.00 € / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

FLI1 Monoclonal antibody specifically detects FLI1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FLI1
Monoclonal
DyLight 350
EWSR2, Friend leukemia integration 1 transcription factor, Friend leukemia virus integration 1, Proto-oncogene Fli-1, SIC-1, Transcription factor ERGB
Recombinant full-length human FLI1 protein
0.1 mL
Cancer, Tumor Suppressors
2313
Store at 4°C in the dark.
IgG κ
Immunohistochemistry (Paraffin)
FLI1/4371R
50 mM Sodium Borate
Rabbit
Protein A purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
FLI1 Antibody (FLI1/4371R), DyLight 350, Novus Biologicals™ > 0.1 mL, DyLight 350

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.