Learn More
Scotch-Brite™ Floor Pads, Red
Product Code. 17168840
Click to view available options
Diameter (Metric):
203 mm
254 mm
280 mm
305 mm
330 mm
355 mm
380 mm
406 mm
432 mm
460 mm
480 mm
505 mm
530 mm
584 mm
missing translation for 'unitSize'
Pack of 10
Pack of 5
Product Code. 17168840
missing translation for 'mfr': Scotch-Brite™ 7100182725
Only null left
Add to basket
Add to basket
Designed for everyday cleaning and light scrubbing to remove light soil, scuff marks, and black heel marks. These pads clean when damp and buff when dry. Open-web construction means this washable, reusable pad resists clogging and loading.
  • For everyday cleaning and light scrubbing
  • Removes light soil, scuff marks, and black heel marks
  • Cleans when damp and buffs when dry
  • Conformable fibers provide maximum pad-to-floor contact
  • Reversible, washable pad resists clogging and loading
  • Consistent, proven performance with long floor pad life
TRUSTED_SUSTAINABILITY
Specifications
Product Type Floor Pad
Color Red
Diameter (Metric) 330 mm
For Use With (Application) Everyday Cleaning, Light Scrubbing