missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ FluoReporter™ Cell-Surface Biotinylation Kit

FluoReporter™ Cell-Surface Biotinylation Kit

Brand:  Molecular Probes™ F20650

375.44 EUR valid until 2024-12-20
Use promo code "21651" to get your promotional price.


Product Code. 10741154

  • 494.00 € / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.

This item is not returnable. View return policy

Description

Description

The FluoReporter™ Cell-Surface Biotinylation Kit provides a convenient method to label proteins exposed on the cell surface including, but not limited to, membrane proteins. The kit supplies 5 vials of biotin-XX SSE, and each vial can be used for several 1-mL labeling reactions (containing 2.5 x 107 cells/mL).

Important Features of FluoReporter™ Protein Labeling Kits:

  • Biotinylation is typically complete in under 1 hour
  • Optimized for 1 mL labeling reactions each containing 2.5 x 107 cells/mL
  • Biotinylated proteins can be subsequently identified using western blot techniques and labeled avidin and streptavidin conjugates (find an anti-biotin antibody using our Secondary Antibody Selector Tool)

    Learn More About Protein Labeling
    We offer a wide selection of Molecular Probes™ protein and antibody labeling kits to fit your starting material and your experimental setup. See Antibody Labeling from A to Z or use our Labeling Chemistry Selection Tool for other choices. To learn more about our various kits read Kits for Labeling Proteins and Nucleic Acids—Section 1.2 in the Molecular Probes™ Handbook.

    For Research Use Only. Not intended for human or animal therapeutic or diagnostic use.
    TRUSTED_SUSTAINABILITY
  • Specifications

    Specifications

    Store in freezer -5°C to -30°C.
    Biotinylation
    Biotin-XX
    1 kit
    Proteins (Cell Surface)
    Biotin-based
    Dye
    FluoReporter™
    Room Temperature
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Promotions

    Promotions

    For Research Use Only. Not for use in diagnostic procedures.