missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Fluorescent Particle Array (Carboxyl, 9 peaks) Kit
Used to develop assays for allergy testing, autoimmune diseases, cardiac markers, cytokine detection, endocrine markers, infectious disease markers, isotyping, genotyping, kinase and phosphorylated protein activity, metabolic markers, and tissue typing
Brand: Novus Biologicals™ NBP3-18593
This item is not returnable.
View return policy
Description

Specifications
Store at 4°C. Do not freeze. | |
Deionized water | |
9 x 1 mL |
Flow Cytometry | |
0.02% Sodium Azide |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novus Biologicals™ Fluorescent Particle Array (Carboxyl, 9 peaks) Kit > 9 x 1 mL
Spot an opportunity for improvement?Share a Content Correction