missing translation for 'onlineSavingsMsg'
Learn More

Hach Lange™ Fluoride Cuvette Test

Brand:  Hach Lange™ LCK323


Product Code. 10215184

  • 121.00 € / Pack of 25
Estimated Shipment: 13-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Order Info

dimRange

Specifications

Specifications

25
15°C to 25°C
Fluoride Cuvette Test
0.1 to 2.5mg/L F
SPADNS
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Fluorid/Fluoride/Fluorure

Signal Word
  • Danger

Hazard Category
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.

Supplemental information
  • MIXTURE LIST-Contains : Hydrochloric acid

SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.