Learn More
Memmert™ Flush-Fit Sets without Air Slots for UF1060
Product Code. 17508805 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
UF1060 Oven
Unit Size:
Each
Product Code. 17508805
Brand: Memmert™ UF1060B29743
Only null left
Add to basket
Add to basket
Stainless steel frame covering gap between oven and wall opening.
  • Stainless steel
  • Without air slots
  • Techincal clarification required
Specifications
For Use With (Equipment) UF1060 Oven
Type Flush-fit Unit
Material Stainless Steel