missing translation for 'onlineSavingsMsg'
Learn More

ZVG Zellstoff Vertrieb™ Folded Towels

Use ZVG Zellstoff Vertrieb™ Folded Towels in either 2-ply or 3-ply folded paper forms. They are available in various sizes, quantities and colors as well.

Brand:  ZVG Zellstoff Vertrieb™ 1542304

 View more versions of this product

Product Code. 10249311

  • 71.35 € / Pack of 3200
Estimated Shipment: 14-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • ZetForm™l paper towel system
  • Various colors
  • Various thicknesses
Specifications

Specifications

Folded Towel
24 cm
24 x 33 cm
33 cm
Paper
2-ply
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
ZVG Zellstoff Vertrieb™ Folded Towels > Dimensions (LxW): 24 x 33cm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.