Learn More
Folic acid, 95-102%, Thermo Scientific Chemicals
30.70 € - 217.00 €
Chemical Identifiers
CAS | 59-30-3 |
---|---|
Molecular Formula | C19H19N7O6 |
Molecular Weight (g/mol) | 441.404 |
MDL Number | MFCD00079305 |
InChI Key | OVBPIULPVIDEAO-LBPRGKRZSA-N |
Synonym | folic acid, folate, pteroylglutamic acid, vitamin m, folacin, folacid, pteglu, vitamin b9, folvite, pteroyl-l-glutamic acid |
PubChem CID | 6037 |
ChEBI | CHEBI:27470 |
IUPAC Name | (2S)-2-[[4-[(2-amino-4-oxo-1H-pteridin-6-yl)methylamino]benzoyl]amino]pentanedioic acid |
SMILES | C1=CC(=CC=C1C(=O)NC(CCC(=O)O)C(=O)O)NCC2=CN=C3C(=N2)C(=O)N=C(N3)N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15435239
|
Thermo Scientific Alfa Aesar
J62937.06 |
5 g |
30.70 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15445239
|
Thermo Scientific Alfa Aesar
J62937.14 |
25 g |
69.90 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15455239
|
Thermo Scientific Alfa Aesar
J62937.22 |
100 g |
217.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Folic acid and its derivatives are essential mediators of one-carbon metabolism within cells.It is especially important in aiding rapid cell division and growth.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
59-30-3 | |
441.404 | |
OVBPIULPVIDEAO-LBPRGKRZSA-N | |
6037 | |
(2S)-2-[[4-[(2-amino-4-oxo-1H-pteridin-6-yl)methylamino]benzoyl]amino]pentanedioic acid |
C19H19N7O6 | |
MFCD00079305 | |
folic acid, folate, pteroylglutamic acid, vitamin m, folacin, folacid, pteglu, vitamin b9, folvite, pteroyl-l-glutamic acid | |
CHEBI:27470 | |
C1=CC(=CC=C1C(=O)NC(CCC(=O)O)C(=O)O)NCC2=CN=C3C(=N2)C(=O)N=C(N3)N |
Specifications
59-30-3 | |
C19H19N7O6 | |
100781 | |
Soluble in water (1.6mg/ml),1 M NaOH (50mg/ml),methanol (slightly),diluted acids,and alkaline solutions. | |
C1=CC(=CC=C1C(=O)NC(CCC(=O)O)C(=O)O)NCC2=CN=C3C(=N2)C(=O)N=C(N3)N | |
441.404 | |
CHEBI:27470 | |
Powder |
Glass bottle | |
MFCD00079305 | |
folic acid, folate, pteroylglutamic acid, vitamin m, folacin, folacid, pteglu, vitamin b9, folvite, pteroyl-l-glutamic acid | |
OVBPIULPVIDEAO-LBPRGKRZSA-N | |
(2S)-2-[[4-[(2-amino-4-oxo-1H-pteridin-6-yl)methylamino]benzoyl]amino]pentanedioic acid | |
6037 | |
441.41 | |
Folic acid |
Safety and Handling
missing translation for 'einecsNumber' : 200-419-0
missing translation for 'rtecsNumber' : LP5425000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only