Learn More
WTW™ Formaldehyde Cell Test Kit
With certificate for optimum precision.
Brand: WTW™ 250406
135.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Convenient and cost-effective
- Assured quality by AQA/IQC
- For infrequent or very large sample throughput in the laboratory
- Sample adding, inserting, measuring and reading
- Reaction cuvette with barcode or method selection
Specifications
Chromotropic acid | |
PhotoLab™ S6, PhotoLab™ S12, PhotoLab™ Spectral photometers | |
25 | |
25 Tests | |
Formaldehyde (HCHO) cell test kit |
For disinfectants, preservatives, applications for cosmetics and chipboard | |
Certificates | |
0.10 - 8.00mg/I HCHO | |
Photometric |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
WTW™ Formaldehyde Cell Test Kit > Cell Test; Detection Range: 0.10 to 8mg/L
Spot an opportunity for improvement?Share a Content Correction