Learn More
Poulten & Graf GmbH FORTUNA™ Burette Bang pattern
Product Code. 10341462
Click to view available options
Capacity (Metric):
2 mL
5 mL
10 mL
Unit Size:
Each
Product Code. 10341462
Brand: Poulten & Graf GmbH 1 03023G
Only null left
Add to basket
Add to basket
Achieve better accuracy and repeatability, especially in titrations. Poulten & Graf GmbH FORTUNA™ Burette Bang pattern delivers accurate and precise volumes.
  • Comformity approved
  • Blue graduation
  • With glass key in upper stopcock
  • With straight glass stopcock
Specifications
Capacity (Metric) 2 mL
Material Borosilicate Glass
Graduations 0.01 mL
For Use With (Application) Burette Bang pattern
Tolerance 0.01 mL
Certifications/Compliance EN ISO 385