Learn More
Watson-Marlow 314X Pumphead
300 Series Pumpheads. Flip top pumpheads for continuous tubing for flow rates to 3 L/min. Extendable up to 6 channels of flow.
Brand:  Watson-Marlow 033.4431.000
352.45 EUR valid until 2024-11-30
Use promo code "21681" to get your promotional price.
Product Code. 12496340
  • 371.00 € / Each
Estimated Shipment: 11-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The 300 series combines performance, practicality and design. With its iconic flip-top design it is an industry standard OEM product in medical device, biotechnology and diagnostic applications, and very popular in many other applications too.
  • Case material: Glass filled polypropylene, IXEF (Polyarylamide), and Stainless steel
  • Rotor material: Electroless nickel plated hardened steel, and Glass filled Nylon
TRUSTED_SUSTAINABILITY
Specifications
Specifications
323 Series Peristaltic Pumps
Polypropylene
4
Pump Extension