missing translation for 'onlineSavingsMsg'
Learn More

Hach Lange™ Free Chlorine Reagent Powder Pillows

Product Code. p-8009395
Click to view available options
Range:
0.02 to 2.0 mg/L Cl2
Unit Size:
Each
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Product Code. 10307182

Brand: Hach Lange™ 2105569

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Hach Lange Free Chlorine Reagent Powder Pillows, Range: 0.02 to 2.00 mg/L Cl2, Certifications/Compliance: EPA compliant, For Use With: Free chlorine determination, All except DR/3000, DR/2000, Parameter Specifications: Free Chlorine, Chlorine Dioxide, Quantity: 100Pack

Specifications

Certifications/Compliance EPA compliant
Volume (Metric) 10 mL
Format Powder
For Use With (Application) Free chlorine determination
For Use With (Equipment) All except DR/3000, DR/2000
Parameter Specifications Free Chlorine, Chlorine Dioxide
Quantity 100 Pack
Range 0.02 to 2.0 mg/L Cl2
Storage Requirements 10°C to 25°C
Test Method 8021, DPD
Type Free Chlorine Reagent Powder Pillows
Show More Show Less
compliance-icons
Product Identifier
  • DPD Free Chlorine Reagent
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P362-Take off contaminated clothing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : di-Sodium hydrogen phosphate, Salt of N,N-Diethyl-p-Phenylenediamine, Ethylenedinitrilotetraacetic
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.