Learn More
BUCHI Freeze Drying Flask Adapter Set
Includes 12 non-standard joint size adapters.
Brand:  BUCHI 11066144
Product Code. 17254674
  • 204.00 € / Each
Estimated Shipment: 25-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With 12 adapters, including filter paper
Specifications
Specifications
Freeze Dryers
26 mm
0.2 kg
12 Adapters, Filter Paper
0.4 lb.