Learn More
Kirsch Philipp™ FrosterMed™ Pharmaceutical Refrigerator
Used as integrated or undercurrent type.
Brand:  Kirsch Philipp™ 10103
Product Code. 10380992
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Includes: One drawer, two shelves
This item is not returnable. View return policy
Description
Description
  • Dynamic ventilated compressor
  • Air discharge via ventilating grille at front
  • Undercounter type
  • Heated door frame
  • Rapid freezing cycle
  • Digital temperature display
  • Key switch
  • Minimum/Maximum temperature memory
  • Optical and acoustic warning device
  • Potential free contact, interface RS485
  • Low-noise compressor
Specifications
Specifications
Laboratory Freezer
-5°C to -25°C
13 to 25 kg
44 x 42 x 47 cm
50/60 Hz
170 kg (gross)
95 L
5 Adjustable
Storage of laboratory chemicals
2.6kWh/24 hr.
160 w
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Kirsch Philipp™ FrosterMed™ Pharmaceutical Refrigerator > FrosterMed Pharmaceutical Refrigerator