missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Memmert™ Full-Sight Glass Door for UN160plus, Max. 250°C
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
UN160plus Oven
Unit Size:
Each
Description
- Accessory
- Full-sight
- Glass (4 layers)
- Temperature range up to 250°C
Specifications
Specifications
| For Use With (Equipment) | UN160plus Oven |
| Temperature (Metric) | 250°C (Max.) |
| Type | Full-sight Door |
| Material | Glass |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction