Learn More
Memmert™ Full-Sight Glass Door for UN260plus, Max. 250°C
Full-sight glass door constructed of four insulated glass layers.
Brand:  Memmert™ UN260plusB0
Product Code. 17559995
  • 872.00 € / Each
Estimated Shipment: 14-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Accessory
  • Full-sight
  • Glass (4 layers)
  • Temperature range up to 250°C
Specifications
Specifications
UN260plus Oven
250°C (Max.)
Full-sight Door
Glass
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ Full-Sight Glass Door for UN260plus, Max. 250°C > For Use With: UN260plus